KEY004181327 Key Organics / BIONET

4-amino-5H,8H-pyrido[2,3-d]pyrimidin-5-one

  • MDL number MFCD03001194
  • Cas Number : 306960-30-5
  • Molecular Weight : 162.152

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 300
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9602

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12R-0218_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart