KEY004181333 Key Organics / BIONET

6-(4-methylphenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde

  • MDL number MFCD03425723
  • Cas Number : 82588-42-9
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167 - 169
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9632

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12R-0276_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart