KEY004181348 Key Organics / BIONET

1-methyl-3-phenyl-1H-pyrazol-5-ol

  • MDL number MFCD03001245
  • Cas Number : 34347-81-4
  • Molecular Weight : 174.203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213 - 215
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64395

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1K-553S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart