KEY004181349 Key Organics / BIONET

5-(aminomethylidene)-2,2-dimethyl-1,3-dioxane-4,6-dione

  • MDL number MFCD01111609
  • Cas Number : 15568-88-4
  • Molecular Weight : 171.152

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210.5 - 211.5
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64579

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1L-411S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart