KEY004181358 Key Organics / BIONET

(2E)-3-{4-[(3-chlorophenyl)methoxy]phenyl}prop-2-enoic acid

  • MDL number MFCD03001246
  • Cas Number : 385383-37-9
  • Molecular Weight : 288.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210 - 212
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65332

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-0028_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart