KEY004181359 Key Organics / BIONET

(2E)-3-{4-[(4-bromophenyl)methoxy]phenyl}prop-2-enoic acid

  • MDL number MFCD03001247
  • Cas Number : 385383-36-8
  • Molecular Weight : 333.181

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225 - 227
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65333

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-0029_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart