KEY004181360 Key Organics / BIONET

(2E)-3-{4-[(2,4-dichlorophenyl)methoxy]phenyl}prop-2-enoic acid

  • MDL number MFCD03001248
  • Cas Number : 385383-41-5
  • Molecular Weight : 323.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234 - 236
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65334

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-0030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart