KEY004181361 Key Organics / BIONET

(2E)-3-{4-[(2-chloro-6-fluorophenyl)methoxy]phenyl}prop-2-enoic acid

  • MDL number MFCD02091011
  • Cas Number : 400825-72-1
  • Molecular Weight : 306.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177 - 179
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65352

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-0049_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart