KEY004181364 Key Organics / BIONET

ethyl 1H,2H-naphtho[2,1-b]furan-2-carboxylate

  • MDL number MFCD03001249
  • Cas Number : 62019-34-5
  • Molecular Weight : 242.274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64 - 66
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65436

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-0193_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart