KEY004181365 Key Organics / BIONET

1H,2H-naphtho[2,1-b]furan-2-carboxylic acid

  • MDL number MFCD03001250
  • Cas Number : 24758-31-4
  • Molecular Weight : 214.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 330
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65440

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-0197_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart