KEY004181373 Key Organics / BIONET

2-acetylbenzonitrile

  • MDL number MFCD00272571
  • Cas Number : 91054-33-0
  • Molecular Weight : 145.161

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46 - 48
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65784

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0269_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart