KEY004181375 Key Organics / BIONET

5-(4-hydroxypiperidin-1-yl)thiophene-2-carbaldehyde

  • MDL number MFCD03425728
  • Cas Number : 207290-72-0
  • Molecular Weight : 211.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 137 - 139
Iupac Name 5-(4-hydroxy-1-piperidinyl)-2-thiophenecarbaldehyde
Inchi Code 1S/C10H13NO2S/c12-7-9-1-2-10(14-9)11-5-3-8(13)4-6-11/h1-2,7-8,13H,3-6H2
Inchi Key QGNLQHONMNFZJP-UHFFFAOYSA-N

Description

Melting Point 137 - 139
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65797

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0289_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart