KEY004181376 Key Organics / BIONET

N-(2,3-dihydro-1H-inden-5-yl)-2-(4-hydroxyphenyl)acetamide

  • MDL number MFCD03425729
  • Cas Number : 321853-28-5
  • Molecular Weight : 267.328

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157 - 158
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65801

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0297_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart