KEY004181377 Key Organics / BIONET

(E)-(ethyl N-[(E)-5-amino-1-phenyl-1H-pyrazole-4-carbonyl]ethanecarbohydrazonate)

  • MDL number MFCD03001206
  • Cas Number : 99347-10-1
  • Molecular Weight : 287.323

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146 - 148
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65804

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0304_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart