KEY004181380 Key Organics / BIONET

[1-methyl-5-(4-methylphenoxy)-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol

  • MDL number MFCD03001207
  • Cas Number : 400073-97-4
  • Molecular Weight : 286.254

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88 - 90
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65816

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0334_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart