KEY004181381 Key Organics / BIONET

1-methyl-5-(4-methylphenoxy)-3-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD03001208
  • Cas Number : 400073-98-5
  • Molecular Weight : 292.338

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90 - 92
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65819

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0339_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart