KEY004181385 Key Organics / BIONET

2-chloro-4-(1H-pyrazol-1-yl)benzoic acid

  • MDL number MFCD03425733
  • Cas Number : 313674-12-3
  • Molecular Weight : 222.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 210 - 213
Iupac Name 2-chloro-4-(1H-pyrazol-1-yl)benzoic acid
Inchi Code 1S/C10H7ClN2O2/c11-9-6-7(13-5-1-4-12-13)2-3-8(9)10(14)15/h1-6H,(H,14,15)
Inchi Key JBLRHBKKTPDCTI-UHFFFAOYSA-N

Description

Melting Point 210 - 213
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65981

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-1487_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart