KEY004181387 Key Organics / BIONET

furan-2-carboxamide

  • MDL number MFCD00236147
  • Cas Number : 609-38-1
  • Molecular Weight : 111.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138 - 140
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66608

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2B-052_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart