KEY004181392 Key Organics / BIONET

[1-methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazol-5-yl]methanol

  • MDL number MFCD01936006
  • Cas Number : 338982-22-2
  • Molecular Weight : 236.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88 - 92
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68231

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2M-320S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart