KEY004181397 Key Organics / BIONET

1-[4-(3-fluoropropoxy)phenyl]ethan-1-one

  • MDL number MFCD03001251
  • Cas Number : 400878-19-5
  • Molecular Weight : 196.221

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68863

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2R-0098_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart