KEY004181402 Key Organics / BIONET

2-(4-methylbenzenesulfonamido)propanoic acid

  • MDL number MFCD00230250
  • Cas Number : 4816-81-3
  • Molecular Weight : 243.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146 - 148
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69371

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0200_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart