KEY004181404 Key Organics / BIONET

methyl 3-amino-4-(1H-1,2,4-triazol-1-yl)benzoate

  • MDL number MFCD03425738
  • Cas Number : 167626-50-8
  • Molecular Weight : 218.216

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139 - 142
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69393

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0238_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart