KEY004181411 Key Organics / BIONET

(E)-(ethyl N-[(E)-5-amino-1-methyl-1H-pyrazole-4-carbonyl]ethanecarbohydrazonate)

  • MDL number MFCD05021852
  • Cas Number : 114936-20-8
  • Molecular Weight : 225.252

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199 - 201
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69427

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0283_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart