KEY004181416 Key Organics / BIONET

2-aminothiophene-3-carboxamide

  • MDL number MFCD00706455
  • Cas Number : 14080-51-4
  • Molecular Weight : 142.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 153 - 155
Iupac Name 2-amino-3-thiophenecarboxamide
Inchi Code 1S/C5H6N2OS/c6-4(8)3-1-2-9-5(3)7/h1-2H,7H2,(H2,6,8)
Inchi Key WHZIZZOTISTHCT-UHFFFAOYSA-N

Description

Melting Point 153 - 155
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69576

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/2T-0905_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart