KEY004181425 Key Organics / BIONET

1-{4-[(2,4-dichlorophenyl)methoxy]phenyl}ethan-1-ol

  • MDL number MFCD03001253
  • Cas Number : 61292-28-2
  • Molecular Weight : 297.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62 - 64
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72509

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0002_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart