KEY004181431 Key Organics / BIONET

3,4-diethyl 5,6-diphenylpyridazine-3,4-dicarboxylate

  • MDL number MFCD03001256
  • Cas Number : 253144-68-2
  • Molecular Weight : 376.412

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114 - 116
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72629

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0184_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart