KEY004181433 Key Organics / BIONET

1-{3-[(2,4-dichlorophenyl)methoxy]phenyl}ethan-1-one

  • MDL number MFCD03001258
  • Cas Number : 400878-28-6
  • Molecular Weight : 295.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71 - 73
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72649

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0215_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart