KEY004181435 Key Organics / BIONET

diphenylpyridazine-3,4-dicarboxylic acid

  • MDL number MFCD03001259
  • Cas Number : 253144-69-3
  • Molecular Weight : 320.304

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140 - 142
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72702

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0293_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart