KEY004181439 Key Organics / BIONET

methyl 1-(4-fluorobenzenesulfonyl)piperidine-4-carboxylate

  • MDL number MFCD02736084
  • Cas Number : 349624-64-2
  • Molecular Weight : 301.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90 - 91
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73002

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-1308_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart