KEY004181440 Key Organics / BIONET

methyl 1-(benzenesulfonyl)piperidine-4-carboxylate

  • MDL number MFCD01045646
  • Cas Number : 311794-15-7
  • Molecular Weight : 283.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83 - 85
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73003

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-1309_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart