KEY004181442 Key Organics / BIONET

7-(benzyloxy)naphthalen-2-ol

  • MDL number MFCD03425751
  • Cas Number : 118495-07-1
  • Molecular Weight : 250.297

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151 - 152
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73046

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0008_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart