KEY004181443 Key Organics / BIONET

3,7,7-trimethyl-4-phenyl-1H,4H,5H,6H,7H,8H,9H-pyrazolo[3,4-b]quinolin-5-one

  • MDL number MFCD03448178
  • Cas Number : 439095-16-6
  • Molecular Weight : 307.397

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 352 - 353
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73047

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0010_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart