KEY004181444 Key Organics / BIONET

4-(furan-3-yl)-3,7,7-trimethyl-1H,4H,5H,6H,7H,8H,9H-pyrazolo[3,4-b]quinolin-5-one

  • MDL number MFCD05021860
  • Cas Number : 439095-18-8
  • Molecular Weight : 297.358

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 296 - 298
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73064

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0040_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart