KEY004181445 Key Organics / BIONET

ethyl 2-{1-[3-(trifluoromethyl)benzoyl]piperidin-4-ylidene}acetate

  • MDL number MFCD03425754
  • Cas Number : 439095-20-2
  • Molecular Weight : 341.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 65 - 67
Iupac Name ethyl {1-[3-(trifluoromethyl)benzoyl]-4-piperidinylidene}acetate
Inchi Code 1S/C17H18F3NO3/c1-2-24-15(22)10-12-6-8-21(9-7-12)16(23)13-4-3-5-14(11-13)17(18,19)20/h3-5,10-11H,2,6-9H2,1H3
Inchi Key MXKDWLPWFAGAPC-UHFFFAOYSA-N

Description

Melting Point 65 - 67
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73069

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0054_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart