KEY004181446 Key Organics / BIONET

7-(diethylamino)-2-oxo-4-(trifluoromethyl)-2H-chromene-3-carboxylic acid

  • MDL number MFCD03425755
  • Cas Number : 333303-14-3
  • Molecular Weight : 329.275

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180 - 182
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73075

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0064_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart