KEY004181447 Key Organics / BIONET

2-(hydroxymethyl)-2-phenyl-1,2,3,4-tetrahydroquinazolin-4-one

  • MDL number MFCD05021861
  • Cas Number : 308832-66-8
  • Molecular Weight : 254.289

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213 - 216
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73079

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0081_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart