KEY004181455 Key Organics / BIONET

4-amino-2-(hydroxymethyl)phenol

  • MDL number MFCD03425760
  • Cas Number : 104333-09-7
  • Molecular Weight : 139.154

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147 - 149
Purity 95
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73123

Safety Information

signalword Warning
pcodes P411,P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0288_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart