KEY004181463 Key Organics / BIONET

[6-(4-bromophenyl)imidazo[2,1-b][1,3]thiazol-5-yl]methanol

  • MDL number MFCD03425764
  • Cas Number : 439095-24-6
  • Molecular Weight : 309.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 197 - 199
Iupac Name [6-(4-bromophenyl)imidazo[2,1-b][1,3]thiazol-5-yl]methanol
Inchi Code 1S/C12H9BrN2OS/c13-9-3-1-8(2-4-9)11-10(7-16)15-5-6-17-12(15)14-11/h1-6,16H,7H2
Inchi Key AKGPVJXKSLVBMY-UHFFFAOYSA-N

Description

Melting Point 197 - 199
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73160

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0339_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart