KEY004181471 Key Organics / BIONET

1-methyl-5-(methylsulfanyl)-1H-1,2,4-triazol-3-amine

  • MDL number MFCD00051817
  • Cas Number : 84827-78-1
  • Molecular Weight : 144.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105 - 107
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74013

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4C-037_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart