KEY004181480 Key Organics / BIONET

2-amino-1-[(4-methoxyphenyl)methyl]-4,5-dimethyl-1H-pyrrole-3-carbonitrile

  • MDL number MFCD03001262
  • Cas Number : 451485-75-9
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142 - 144
Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76214

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4P-315S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart