KEY004181485 Key Organics / BIONET

methyl (2E)-3-[(E)-4-(trifluoromethyl)benzoyloxy]prop-2-enoate

  • MDL number MFCD03001265
  • Cas Number : 400878-21-9
  • Molecular Weight : 274.195

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86 - 88
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76500

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-0307_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart