KEY004181486 Key Organics / BIONET

methyl (2E)-3-[(E)-3,4,5-trimethoxybenzoyloxy]prop-2-enoate

  • MDL number MFCD03001266
  • Cas Number : 400878-22-0
  • Molecular Weight : 296.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 109 - 111
Iupac Name (1E)-3-methoxy-3-oxo-1-propenyl 3,4,5-trimethoxybenzoate
Inchi Code 1S/C14H16O7/c1-17-10-7-9(8-11(18-2)13(10)20-4)14(16)21-6-5-12(15)19-3/h5-8H,1-4H3/b6-5+
Inchi Key WBHVFAMRSLOODA-AATRIKPKSA-N

Description

Melting Point 109 - 111
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76513

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4R-0326_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart