KEY004181487 Key Organics / BIONET

methyl (2E)-3-[(E)-4-aminobenzoyloxy]prop-2-enoate

  • MDL number MFCD03001267
  • Cas Number : 400878-23-1
  • Molecular Weight : 221.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 186 - 188
Iupac Name (1E)-3-methoxy-3-oxo-1-propenyl 4-aminobenzoate
Inchi Code 1S/C11H11NO4/c1-15-10(13)6-7-16-11(14)8-2-4-9(12)5-3-8/h2-7H,12H2,1H3/b7-6+
Inchi Key MIHJXRPOCKDWIE-VOTSOKGWSA-N

Description

Melting Point 186 - 188
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76514

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4R-0328_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart