KEY004181489 Key Organics / BIONET

1-(4-chlorobenzenesulfonyl)piperidine-4-carboxylic acid

  • MDL number MFCD00723388
  • Cas Number : 314744-43-9
  • Molecular Weight : 303.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206 - 208
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76656

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-1137_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart