KEY004181494 Key Organics / BIONET

1-(4-methoxybenzenesulfonyl)piperidine-4-carboxylic acid

  • MDL number MFCD02331441
  • Cas Number : 385398-15-2
  • Molecular Weight : 299.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163 - 165
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76726

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-1334_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart