KEY004181503 Key Organics / BIONET

ethyl 5-amino-1-(2,4-dichlorophenyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD01569432
  • Cas Number : 83279-66-7
  • Molecular Weight : 300.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 175 - 178
Iupac Name ethyl 5-amino-1-(2,4-dichlorophenyl)-1H-pyrazole-4-carboxylate
Inchi Code 1S/C12H11Cl2N3O2/c1-2-19-12(18)8-6-16-17(11(8)15)10-4-3-7(13)5-9(10)14/h3-6H,2,15H2,1H3
Inchi Key XOBRIYKLMQJCBQ-UHFFFAOYSA-N

Description

Melting Point 175 - 178
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76809

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4T-0237_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart