KEY004181507 Key Organics / BIONET

2,4-dichloro-5-[(3-fluorophenyl)methoxy]aniline

  • MDL number MFCD03425777
  • Cas Number : 439095-33-7
  • Molecular Weight : 286.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84 - 86
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76843

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0294_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart