KEY004181508 Key Organics / BIONET

3-(2H-1,3-benzodioxol-5-yloxy)butan-2-one

  • MDL number MFCD03425778
  • Cas Number : 23973-82-2
  • Molecular Weight : 208.213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 30 - 32
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76862

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0317_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart