KEY004181524 Key Organics / BIONET

4-(2,4-dichlorophenyl)-1,3-thiazol-2-amine

  • MDL number MFCD01114990
  • Cas Number : 93209-97-3
  • Molecular Weight : 245.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 156 - 158
Iupac Name 4-(2,4-dichlorophenyl)-1,3-thiazol-2-ylamine
Inchi Code 1S/C9H6Cl2N2S/c10-5-1-2-6(7(11)3-5)8-4-14-9(12)13-8/h1-4H,(H2,12,13)
Inchi Key APJACVDBTHESJL-UHFFFAOYSA-N

Description

Melting Point 156 - 158
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79045

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5K-651S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart