KEY004181527 Key Organics / BIONET

2-methanesulfinylbenzoic acid

  • MDL number MFCD01936003
  • Cas Number : 19093-34-6
  • Molecular Weight : 184.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 166 - 170
Iupac Name 2-(methylsulfinyl)benzoic acid
Inchi Code 1S/C8H8O3S/c1-12(11)7-5-3-2-4-6(7)8(9)10/h2-5H,1H3,(H,9,10)
Inchi Key JVYFTEBQXOBPAB-UHFFFAOYSA-N

Description

Melting Point 166 - 170
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79261

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5L-554S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart